Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166663.1 | NP_001160135.1 | 2 | 361 | |
NM_001166664.1 | NP_001160136.1 | 2 | 361 | ||
NM_016382.3 | NP_057466.1 | 2 | 361 | ||
XM_011509621.2 | XP_011507923.1 | 2 | 346 | ||
XM_011509622.2 | XP_011507924.1 | 2 | 346 | ||
GenBank mRNA | AF105261.1 | - | 2 | 178 | |
AF107761.2 | - | 2 | 244 | ||
AF117711.1 | - | 2 | 315 | ||
AF145782.1 | - | 2 | 136 | ||
AF242540.1 | - | 2 | 219 | ||
AJ245375.1 | - | 2 | 159 | ||
AJ245376.1 | - | 2 | 159 | ||
AJ245377.1 | - | 2 | 159 | ||
AK295950.1 | - | 2 | 346 | ||
AK313201.1 | - | 2 | 282 | ||
BC028073.1 | - | 2 | 361 | 5213370 | |
BC041607.1 | - | 2 | 163 | ||
BC053985.1 | - | 2 | 147 | 5484877 |