Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000860.5 | NP_000851.2 | 5 | 899 | |
NM_001145816.2 | NP_001139288.1 | 5 | 899 | ||
NM_001256301.1 | NP_001243230.1 | 5 | 629 | ||
NM_001256306.1 | NP_001243235.1 | 3 | 695 | ||
NM_001256307.1 | NP_001243236.1 | 4 | 792 | ||
GenBank mRNA | AK058013.1 | - | 5 | 488 | |
AK300125.1 | - | 3 | 285 | ||
AK300524.1 | - | 4 | 535 | ||
AK300940.1 | - | 5 | 488 | ||
AK314624.1 | - | 5 | 488 | ||
BC018986.2 | - | 5 | 461 | 3638799 | |
J05594.1 | - | 5 | 469 | ||
L76465.1 | - | 5 | 469 | ||
X82460.1 | - | 5 | 452 |