Gene Transcripts

Gene Symbol
GMPR
Entrez Gene ID
2766
Gene Name
guanosine monophosphate reductase
Gene Aliases
GMPR 1, GMPR1
Chromosome Location
Chr. 6: 16238580 - 16295549 on Build GRCh38
UniGene ID
Hs.484741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006877.3 NP_006868.3 2 222
XM_011514508.2 XP_011512810.1 2 210
GenBank mRNA BC008281.1 - 2 222
M24470.1 - 2 179

View Full Product Details