Gene Transcripts

Gene Symbol
HVCN1
Entrez Gene ID
84329
Gene Name
hydrogen voltage gated channel 1
Gene Aliases
HV1, VSOP
Chromosome Location
Chr. 12: 110648686 - 110704945 on Build GRCh38
UniGene ID
Hs.211511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040107.1 NP_001035196.1 7 865
NM_001256413.1 NP_001243342.1 6 825
NM_032369.3 NP_115745.2 7 856
XM_005253948.2 XP_005254005.1 7 943
XM_011538838.1 XP_011537140.1 8 1281
XM_011538839.2 XP_011537141.1 8 1015
XM_011538840.2 XP_011537142.1 8 1047
XM_011538841.2 XP_011537143.1 8 1025
XM_011538842.2 XP_011537144.1 8 1005
XM_011538844.2 XP_011537146.1 8 1020
XM_011538845.1 XP_011537147.1 7 900
XM_011538846.2 XP_011537148.1 8 974
XM_011538847.1 XP_011537149.1 7 875
XM_017020026.1 XP_016875515.1 8 989
XM_017020027.1 XP_016875516.1 7 874
GenBank mRNA AK293543.1 - 6 791
AY359054.1 - 7 884
BC007277.1 - 7 1308 3343102
BC009731.1 - 7 807 3877292
BC032672.2 - 6 1429 5577070
GU211002.1 - 5 675

View Full Product Details