Gene Transcripts
- Gene Symbol
- CDK1
- Entrez Gene ID
- 983
- Gene Name
- cyclin dependent kinase 1
- Gene Aliases
- CDC2, CDC28A, P34CDC2
- Chromosome Location
- Chr. 10: 60778331 - 60794852 on Build GRCh38
- UniGene ID
- Hs.732435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320918.1 |
NP_001307847.1
|
6, 7 |
817 |
|
NM_001786.4 |
NP_001777.1
|
6, 7 |
805 |
|
NM_033379.4 |
NP_203698.1
|
5, 6 |
634 |
|
XM_005270303.3 |
XP_005270360.1
|
6, 7 |
887 |
|
GenBank mRNA |
AK291939.1 |
- |
6, 7 |
742 |
|
AW976143.1 |
- |
5, 6 |
549 |
|
BC014563.1 |
- |
5, 6 |
676 |
2967728 |
BC107750.1 |
- |
7 |
791 |
6710672 |
BT007004.1 |
- |
5, 6 |
663 |
|
CR933728.1 |
- |
5, 6 |
3032 |
|
D88357.1 |
- |
4, 5 |
519 |
|
X05360.1 |
- |
6, 7 |
789 |
|
Y00272.1 |
- |
6, 7 |
789 |
|
View Full Product Details