Gene Transcripts

Gene Symbol
CEBPA-AS1
Entrez Gene ID
80054
Gene Name
CEBPA antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 19: 33302857 - 33305057 on Build GRCh38
UniGene ID
Hs.287521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026887.1 1 1148
GenBank mRNA AK022417.1 - 1 1050
BC027959.1 - 1 1134
BC113872.1 - 1 935
BC114482.1 - 1 935

View Full Product Details