Gene Transcripts

Gene Symbol
ZDHHC6
Entrez Gene ID
64429
Gene Name
zinc finger DHHC-type containing 6
Gene Aliases
DHHC-6, ZNF376
Chromosome Location
Chr. 10: 112424411 - 112448335 on Build GRCh38
UniGene ID
Hs.196990
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303134.1 NP_001290063.1 3 781
NM_022494.2 NP_071939.1 3 793
XM_005270051.4 XP_005270108.1 3 1728
XM_006717948.3 XP_006718011.1 3 899
XM_006717949.3 XP_006718012.1 3 790
XM_011540091.2 XP_011538393.1 3 781
XM_011540092.2 XP_011538394.1 3 790
XM_011540093.1 XP_011538395.1 3 899
XM_011540094.2 XP_011538396.1 3 758
XM_011540095.2 XP_011538397.1 3 770
XM_017016561.1 XP_016872050.1 3 719
XM_017016562.1 XP_016872051.1 3 887
XM_017016563.1 XP_016872052.1 3 700
XM_017016564.1 XP_016872053.1 3 899
XM_017016565.1 XP_016872054.1 3 965
XM_017016566.1 XP_016872055.1 3 778
XM_017016567.1 XP_016872056.1 3 778
XM_017016568.1 XP_016872057.1 3 696
GenBank mRNA AF267740.1 - 3 747
AK025605.1 - 3 748
AK223086.1 - 3 747
AK308026.1 - 3 941
BC007213.1 - 3 723 2960984
BC017434.2 - 3 723 2960984
BX647270.1 - 2 2221

View Full Product Details