Gene Transcripts

Gene Symbol
TSSK2
Entrez Gene ID
23617
Gene Name
testis specific serine kinase 2
Gene Aliases
DGS-G, SPOGA2, STK22B, TSK2
Chromosome Location
Chr. 22: 19130808 - 19132623 on Build GRCh38
UniGene ID
Hs.694070
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053006.4 NP_443732.3 1 1744
GenBank mRNA AF362953.1 - 1 1224
BC037781.2 - 1 1275 5273397
L77564.1 - 1 1744

View Full Product Details