Gene Transcripts
- Gene Symbol
- PTP4A2
- Entrez Gene ID
- 8073
- Gene Name
- protein tyrosine phosphatase type IVA, member 2
- Gene Aliases
- HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b
- Chromosome Location
- Chr. 1: 31906421 - 31942635 on Build GRCh38
- UniGene ID
- Hs.470477
- Species
- Homo sapiens
- Gene Symbol
- KIF23
- Entrez Gene ID
- 9493
- Gene Name
- kinesin family member 23
- Gene Aliases
- CHO1, KNSL5, MKLP-1, MKLP1
- Chromosome Location
- Chr. 15: 69414246 - 69448427 on Build GRCh38
- UniGene ID
- Hs.270845
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X67155.2 |
- |
|
20 |
|
View Full Product Details