Gene Transcripts

Gene Symbol
MAPK11
Entrez Gene ID
5600
Gene Name
mitogen-activated protein kinase 11
Gene Aliases
P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta
Chromosome Location
Chr. 22: 50263713 - 50270393 on Build GRCh38
UniGene ID
Hs.57732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002751.6 NP_002742.3 4 481
NR_110887.1 4 481
XM_011530706.1 XP_011529008.1 2 205
GenBank mRNA AF001008.1 - 4 380
AF001174.1 - 4 386
AF031135.1 - 4 395
AK291845.1 - 4 432
AK299745.1 - 3 393
AK308969.1 - 4 397
AK309046.1 - 4 583
BC027933.1 - 4 438 5199628
BX361670.2 - 4 465
BX444192.2 - 4 480
CR456514.1 - 4 382
U53442.1 - 4 423
U92268.1 - 4 384
Y14440.1 - 4 386

View Full Product Details