Gene Transcripts

Gene Symbol
ODF2
Entrez Gene ID
4957
Gene Name
outer dense fiber of sperm tails 2
Gene Aliases
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location
Chr. 9: 128455155 - 128501292 on Build GRCh38
UniGene ID
Hs.129055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242352.1 NP_001229281.1 13, 14 1781
NM_001242353.1 NP_001229282.1 13, 14 1867
NM_001242354.1 NP_001229283.1 12, 13 1553
NM_002540.4 NP_002531.3 15, 16 2069
NM_153432.1 NP_702910.1 12, 13 1627
NM_153433.1 NP_702911.1 13, 14 1794
NM_153435.1 NP_702913.1 13, 14 1687
NM_153436.1 NP_702914.1 12, 13 1451
NM_153437.2 NP_702915.1 13, 14 1717
NM_153439.1 NP_702917.1 12, 13 1570
NM_153440.1 NP_702918.1 12, 13 1497
XM_005252010.2 XP_005252067.1 12, 13 1579
XM_005252011.2 XP_005252068.1 12, 13 1519
XM_005252012.3 XP_005252069.1 13, 14 1769
XM_005252013.3 XP_005252070.1 13, 14 1716
XM_005252014.3 XP_005252071.1 13, 14 1765
XM_005252017.4 XP_005252074.1 13, 14 1670
XM_005252018.2 XP_005252075.1 14, 15 1875
XM_005252019.3 XP_005252076.1 14, 15 1894
XM_005252022.2 XP_005252079.1 13, 14 1693
XM_005252023.2 XP_005252080.1 13, 14 1664
XM_005252024.3 XP_005252081.1 13, 14 1683
XM_005252026.4 XP_005252083.1 13, 14 1612
XM_005252029.2 XP_005252086.1 13, 14 1693
XM_006717126.3 XP_006717189.1 13, 14 1769
XM_011518722.2 XP_011517024.1 13, 14 1693
XM_011518723.2 XP_011517025.1 13, 14 1636
XM_011518724.2 XP_011517026.1 14, 15 1857
XM_011518725.2 XP_011517027.1 14, 15 1861
XM_011518726.2 XP_011517028.1 14, 15 1879
XM_011518728.2 XP_011517030.1 13, 14 1630
XM_011518729.2 XP_011517031.1 13, 14 1566
XM_011518730.2 XP_011517032.1 13, 14 1693
XM_011518731.2 XP_011517033.1 12, 13 1522
XM_011518732.2 XP_011517034.1 13, 14 1479
XM_011518733.2 XP_011517035.1 12, 13 1465
XM_011518744.1 XP_011517046.1 12, 13 1519
XM_017014764.1 XP_016870253.1 14, 15 1850
XM_017014765.1 XP_016870254.1 14, 15 1869
XM_017014766.1 XP_016870255.1 12, 13 1579
XM_017014767.1 XP_016870256.1 14, 15 1861
XM_017014768.1 XP_016870257.1 13, 14 1653
XM_017014769.1 XP_016870258.1 13, 14 1682
XM_017014770.1 XP_016870259.1 14, 15 1837
XM_017014771.1 XP_016870260.1 14, 15 1818
XM_017014772.1 XP_016870261.1 14, 15 1894
XM_017014773.1 XP_016870262.1 14, 15 1870
XM_017014774.1 XP_016870263.1 13, 14 1580
XM_017014775.1 XP_016870264.1 14, 15 1837
XM_017014776.1 XP_016870265.1 14, 15 1818
XM_017014777.1 XP_016870266.1 13, 14 1620
XM_017014778.1 XP_016870267.1 13, 14 1723
XM_017014779.1 XP_016870268.1 12, 13 1464
XM_017014780.1 XP_016870269.1 13, 14 1647
XM_017014781.1 XP_016870270.1 12, 13 1402
GenBank mRNA AF012549.1 - 13, 14 1657
AF053970.1 - 11, 12 1356
AK093367.1 - 12, 13 1577
AK299303.1 - 13, 14 1629
AK300793.1 - 12, 13 1402
AK301754.1 - 12, 13 1494
AK301842.1 - 12, 13 1419
AK301855.1 - 12, 13 1473
AK302684.1 - 13, 14 1721
AY319414.1 - 12, 13 1603
AY366499.1 - 13, 14 1564
BC010629.1 - 15, 16 1867 3874501
BC091500.1 - 13, 14 1681 30378678
DQ444713.1 - 11, 12 1394
DQ444714.1 - 11, 12 1337

View Full Product Details