Gene Transcripts

Gene Symbol
HNRNPA1
Entrez Gene ID
3178
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
Chromosome Location
Chr. 12: 54280690 - 54287088 on Build GRCh38
UniGene ID
Hs.546261
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002136.3 NP_002127.1 8, 9 1021
NM_031157.3 NP_112420.1 9, 10 1177
NR_135167.1 8, 9 1021
XM_005268826.1 XP_005268883.1 9, 10 1171
XM_017019251.1 XP_016874740.1 7, 8 940
GenBank mRNA AK129814.1 - 8, 9 963
AK291113.1 - 8, 9 961
AK298176.1 - 5, 6 523
AK303303.1 - 8, 9 765
AK308679.1 - 7, 8 915
BC002355.2 - 8, 9 942 2821751
BC004945.1 - 1 838
BC009600.1 - 8, 9 964 4134638
BC012158.1 - 8, 9 924 4564655
BC020442.2 - 1 931
BC033714.1 - 8, 9 953 4420039
BC035253.1 - 4, 5 380
BC052296.1 - 8, 9 933 6297642
BC070315.1 - 8, 9 963 4101267
BC071945.1 - 1 953 5475712
BC073162.1 - 1 947 6154972
BC074502.1 - 8, 9 946 3455742
BC103707.1 - 7, 8 772 6154800
BC104236.1 - 5, 6 445
BC104237.1 - 5, 6 445
BU501222.1 - 1 394
U00947.1 - 8, 9 911
X04347.1 - 5, 6 531
X06747.1 - 8, 9 988
X79536.1 - 8, 9 929

View Full Product Details