Gene Transcripts

Gene Symbol
AADACL2
Entrez Gene ID
344752
Gene Name
arylacetamide deacetylase like 2
Gene Aliases
Chromosome Location
Chr. 3: 151733916 - 151758016 on Build GRCh38
UniGene ID
Hs.144710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207365.3 NP_997248.2 2 439
XM_011512753.1 XP_011511055.1 2 467
GenBank mRNA BC065724.1 - 2 432
CU449274.1 - 2 261

View Full Product Details