Gene Transcripts

Gene Symbol
Pigr
Entrez Gene ID
18703
Gene Name
polymeric immunoglobulin receptor
Gene Aliases
Chromosome Location
Chr. 1: 130826684 - 130852249 on Build GRCm38
UniGene ID
Mm.276414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011082.3 NP_035212.2 4 777
GenBank mRNA AK128944.1 - 4 777
AK142656.1 - 4 779
BC013556.1 - 4 710 3601095
U06431.1 - 4 681

View Full Product Details