Gene Transcripts

Gene Symbol
Eml6
Entrez Gene ID
237711
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
2900083P10Rik, C230094A16Rik, EMAP-6
Chromosome Location
Chr. 11: 29742586 - 30026034 on Build GRCm38
UniGene ID
Mm.445271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146016.2 NP_666128.2 37 5975
GenBank mRNA AK081418.1 - 2 326
BC024726.1 - 9 1410 5362808

View Full Product Details