Gene Transcripts

Gene Symbol
Nfkbib
Entrez Gene ID
18036
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta
Gene Aliases
I(Kappa)B(beta), I-kappa-B-beta, IKB-beta, IKappaBbeta, IkB, IkBb, NF-kappa-BIB, ikB-B
Chromosome Location
Chr. 7: 28758251 - 28766639 on Build GRCm38
UniGene ID
Mm.220333
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK010218.1 - 3, 4 666
AK038631.1 - 3, 4 714
AK156731.1 - 3, 4 640
AY176058.1 - 3, 4 673
AY176059.1 - 3, 4 704
BC021938.1 - 3, 4 671 5136809
U19799.1 - 3, 4 685

View Full Product Details