Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009968.3 | NP_034098.1 | 9 | 1663 | |
GenBank mRNA | AK098082.1 | - | 9 | 1147 | |
AK133820.1 | - | 10 | 1763 | ||
AK134281.1 | - | 9 | 1148 | ||
BC043076.1 | - | 9 | 1124 | 6404996 | |
D78646.1 | - | 6 | 690 | ||
S70056.1 | - | 9 | 1175 |