Gene Transcripts
- Gene Symbol
- Apol7c
- Entrez Gene ID
- 108956
- Gene Name
- apolipoprotein L 7c
- Gene Aliases
- 2210421G13Rik
- Chromosome Location
- Chr. 15: 77524852 - 77533335 on Build GRCm38
- UniGene ID
- Mm.86391
- Species
- Mus musculus
- Gene Symbol
- Apol7a
- Entrez Gene ID
- 75761
- Gene Name
- apolipoprotein L 7a
- Gene Aliases
- 9130022K13Rik, Apol3
- Chromosome Location
- Chr. 15: 77388217 - 77399110 on Build GRCm38
- UniGene ID
- Mm.232636
- Species
- Mus musculus
- Gene Symbol
- Gm8221
- Entrez Gene ID
- 666661
- Gene Name
- apolipoprotein L 7c pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 77618968 - 77627398 on Build GRCm38
- UniGene ID
- Mm.390818
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052510.1 |
- |
1 |
668 |
|
View Full Product Details