Gene Transcripts

Gene Symbol
PAX9
Entrez Gene ID
5083
Gene Name
paired box 9
Gene Aliases
STHAG3
Chromosome Location
Chr. 14: 36657568 - 36677807 on Build GRCh38
UniGene ID
Hs.132576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006194.3 NP_006185.1 3 1145
GenBank mRNA AB248958.1 - 2 588
AK313627.1 - 2 600
BC001159.1 - 2 608 3534582
U59628.1 - 1 279
X92850.1 - 1 395

View Full Product Details