Gene Transcripts

Gene Symbol
Top1
Entrez Gene ID
21969
Gene Name
topoisomerase (DNA) I
Gene Aliases
AI467334, D130064I21Rik, Top-1
Chromosome Location
Chr. 2: 160645897 - 160722764 on Build GRCm38
UniGene ID
Mm.217233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009408.2 NP_033434.2 10 1145
GenBank mRNA AK145538.1 - 10 1155
D10061.1 - 10 812
L20632.1 - 10 809

View Full Product Details