Gene Transcripts

Gene Symbol
Slc22a8
Entrez Gene ID
83500
Gene Name
solute carrier family 22 (organic anion transporter), member 8
Gene Aliases
OCT3, Oat3, Roct
Chromosome Location
Chr. 1: 224800080 - 224818482 on Build Rnor_6.0
UniGene ID
Rn.23348
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031332.1 NP_112622.1 2 230
GenBank mRNA AB017446.1 - 2 230
BC081777.1 - 2 228 7109603
FQ209755.1 - 242

View Full Product Details