Gene Transcripts

Gene Symbol
F2rl3
Entrez Gene ID
116498
Gene Name
coagulation factor II (thrombin) receptor-like 3
Gene Aliases
PAR4
Chromosome Location
Chr. 16: 18817797 - 18819790 on Build Rnor_6.0
UniGene ID
Rn.81079
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053808.1 NP_446260.1 2 1252
GenBank mRNA AF269246.2 - 2 1252
AF310216.1 - 2 1280

View Full Product Details