Gene Transcripts

Gene Symbol
Casp4
Entrez Gene ID
114555
Gene Name
caspase 4, apoptosis-related cysteine peptidase
Gene Aliases
Casp11
Chromosome Location
Chr. 8: 2616391 - 2651682 on Build Rnor_6.0
UniGene ID
Rn.16195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053736.2 NP_446188.2 2 328
GenBank mRNA AY029283.1 - 2 351
BC104720.1 - 2 328 7382967
FQ227402.1 - 2 365
FQ232279.1 - 2 365

View Full Product Details