Gene Transcripts

Gene Symbol
Lhx1
Entrez Gene ID
257634
Gene Name
LIM homeobox 1
Gene Aliases
Chromosome Location
Chr. 10: 71843991 - 71849293 on Build Rnor_6.0
UniGene ID
Rn.127825
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145880.3 NP_665887.3 3 1091
GenBank mRNA FQ213041.1 - 2 361
S71523.1 - 3 1091

View Full Product Details