Gene Transcripts

Gene Symbol
Vamp3
Entrez Gene ID
29528
Gene Name
vesicle-associated membrane protein 3
Gene Aliases
Chromosome Location
Chr. 5: 168126157 - 168136628 on Build Rnor_6.0
UniGene ID
Rn.219999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057097.2 NP_476438.1 5 403
GenBank mRNA BC088119.1 - 5 403
S63830.1 - 455

View Full Product Details