Gene Transcripts

Gene Symbol
Slc9a2
Entrez Gene ID
24783
Gene Name
solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2
Gene Aliases
Nhe2
Chromosome Location
Chr. 9: 47386581 - 47491813 on Build Rnor_6.0
UniGene ID
Rn.11047
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113335.1 NP_001106806.1 2 899
NM_012653.2 NP_036785.2 1 896
GenBank mRNA L11004.1 - 1 911
L11236.1 - 2 899

View Full Product Details