Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114121.2 | NP_001107593.1 | 13 | 2283 | |
NM_001114122.2 | NP_001107594.1 | 13 | 2283 | ||
NM_001244846.1 | NP_001231775.1 | 12 | 2181 | ||
NM_001274.5 | NP_001265.2 | 13 | 1626 | ||
NR_045204.1 | 12 | 2059 | |||
NR_045205.1 | 13 | 1820 | |||
XM_011542560.2 | XP_011540862.1 | 12 | 2285 | ||
XM_011542561.2 | XP_011540863.1 | 12 | 2285 | ||
XM_011542562.2 | XP_011540864.1 | 12 | 1535 | ||
XM_011542563.2 | XP_011540865.1 | 12 | 1600 | ||
XM_017017146.1 | XP_016872635.1 | 13 | 1470 | ||
GenBank mRNA | AB451222.1 | - | 12 | 1386 | |
AB451345.1 | - | 12 | 1386 | ||
AF016582.1 | - | 12 | 1420 | ||
AF032874.1 | - | 12 | 1386 | ||
AK292549.1 | - | 13 | 1820 | ||
AK293143.1 | - | 12 | 1346 | ||
AK299783.1 | - | 12 | 1220 | ||
AK300082.1 | - | 12 | 1469 | ||
BC004202.2 | - | 13 | 1445 | 3530606 | |
BC017575.1 | - | 13 | 1544 | 4091997 | |
BG191840.1 | - | 1 | 53 | ||
BG216281.1 | - | 1 | 50 | ||
BG218896.1 | - | 2 | 157 | ||
JF289264.1 | - | 12 | 1261 |