Gene Transcripts
- Gene Symbol
- HDAC2
- Entrez Gene ID
- 3066
- Gene Name
- histone deacetylase 2
- Gene Aliases
- HD2, RPD3, YAF1
- Chromosome Location
- Chr. 6: 113936156 - 113971195 on Build GRCh38
- UniGene ID
- Hs.3352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001527.3 |
NP_001518.3
|
6, 7 |
923 |
|
NR_033441.1 |
|
7, 8 |
951 |
|
NR_073443.1 |
|
6, 7 |
829 |
|
XM_011535788.1 |
XP_011534090.1
|
7, 8 |
675 |
|
XM_017010799.1 |
XP_016866288.1
|
7, 8 |
897 |
|
GenBank mRNA |
AB209190.1 |
- |
2, 3 |
729 |
|
AK092156.1 |
- |
6, 7 |
829 |
|
AK097376.1 |
- |
7, 8 |
885 |
|
AK296856.1 |
- |
6, 7 |
923 |
|
BC031055.1 |
- |
6, 7 |
887 |
|
U31814.1 |
- |
6, 7 |
840 |
|
View Full Product Details