Gene Transcripts
- Gene Symbol
- AURKA
- Entrez Gene ID
- 6790
- Gene Name
- aurora kinase A
- Gene Aliases
- AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
- Chromosome Location
- Chr. 20: 56369389 - 56392337 on Build GRCh38
- UniGene ID
- Hs.250822
- Species
- Homo sapiens
- Gene Symbol
- AURKAPS1
- Entrez Gene ID
- 6791
- Gene Name
- aurora kinase A pseudogene 1
- Gene Aliases
- AurAps1, STK6P
- Chromosome Location
- Chr. 1: 220266179 - 220267715 on Build GRCh38
- UniGene ID
- Hs.654849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001587.1 |
|
1 |
1230 |
|
View Full Product Details