Gene Transcripts

Gene Symbol
CCL15-CCL14
Entrez Gene ID
348249
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 17: 35983656 - 36002038 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027921.3 2 644
NR_027922.3 2 644
GenBank mRNA BC140941.1 - 2 227

Gene Symbol
CCL15
Entrez Gene ID
6359
Gene Name
C-C motif chemokine ligand 15
Gene Aliases
HCC-2, HMRP-2B, LKN-1, LKN1, MIP-1 delta, MIP-1D, MIP-5, MRP-2B, NCC-3, NCC3, SCYA15, SCYL3, SY15
Chromosome Location
Chr. 17: 35997582 - 36002038 on Build GRCh38
UniGene ID
Hs.272493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032965.5 NP_116741.2 2 644
GenBank mRNA AF031587.1 - 2 98
BX098368.1 - 2 644
BX497188.1 - 2 112
U58914.1 - 2 158

View Full Product Details