Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282644.1 NP_001269573.1 16, 17 1927
NM_001282645.1 NP_001269574.1 15, 16 1868
NM_001323265.1 NP_001310194.1 16, 17 1984
NM_001323266.1 NP_001310195.1 15, 16 1866
NM_001323267.1 NP_001310196.1 16, 17 1927
NM_006257.4 NP_006248.1 16, 17 1929
XM_005252496.4 XP_005252553.1 16, 17 2029
XM_005252497.4 XP_005252554.1 16, 17 2031
XM_006717465.3 XP_006717528.1 18, 19 4424
XM_017016410.1 XP_016871899.1 17, 18 4335
XM_017016411.1 XP_016871900.1 17, 18 4363
GenBank mRNA AK293935.1 - 15, 16 1806
AK308099.1 - 16, 17 1902
AK312411.1 - 16, 17 1880
BC101465.1 - 15, 16 1848 8068971
BC113359.1 - 15, 16 1848 8322411
BX647657.1 - 15, 16 2014
L01087.1 - 16, 17 1923
L07032.1 - 15, 16 1850

View Full Product Details