Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 887 | |
GenBank mRNA | AB449912.1 | - | 6 | 776 | |
AK293521.1 | - | 5 | 717 | ||
AK293875.1 | - | 6 | 846 | ||
AK294776.1 | - | 6 | 785 | ||
AK297924.1 | - | 3 | 292 | ||
AK298884.1 | - | 5 | 746 | ||
AK316199.1 | - | 4 | 567 | ||
BC036942.1 | - | 3 | 316 | ||
BC052609.1 | - | 5 | 677 | ||
BC082255.1 | - | 4 | 787 | ||
BC101817.1 | - | 6 | 813 | 8069323 | |
BC101819.1 | - | 6 | 813 | 8069325 | |
BC125088.1 | - | 6 | 802 | 40117866 | |
BC125089.1 | - | 5 | 647 | ||
L08240.1 | - | 5 | 666 |