Gene Transcripts

Gene Symbol
NOBOX
Entrez Gene ID
135935
Gene Name
NOBOX oogenesis homeobox
Gene Aliases
OG-2, OG2, OG2X, POF5, TCAG_12042
Chromosome Location
Chr. 7: 144396805 - 144410438 on Build GRCh38
UniGene ID
Hs.558628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080413.3 NP_001073882.3 4 823
XM_017011742.1 XP_016867231.1 4 1034

View Full Product Details