Gene Transcripts

Gene Symbol
DBX1
Entrez Gene ID
120237
Gene Name
developing brain homeobox 1
Gene Aliases
Chromosome Location
Chr. 11: 20156214 - 20160324 on Build GRCh38
UniGene ID
Hs.558604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029865.2 NP_001025036.2 4 972

View Full Product Details