Gene Transcripts
- Gene Symbol
- KIF14
- Entrez Gene ID
- 9928
- Gene Name
- kinesin family member 14
- Gene Aliases
- MKS12
- Chromosome Location
- Chr. 1: 200551497 - 200620791 on Build GRCh38
- UniGene ID
- Hs.3104
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014875.2 |
NP_055690.1
|
4, 5 |
1902 |
|
XM_011510231.2 |
XP_011508533.1
|
5, 6 |
1996 |
|
XM_011510232.2 |
XP_011508534.1
|
4, 5 |
1658 |
|
XM_011510233.2 |
XP_011508535.1
|
5, 6 |
1789 |
|
XM_011510235.2 |
XP_011508537.1
|
5, 6 |
1501 |
|
XM_017003005.1 |
XP_016858494.1
|
3, 4 |
2079 |
|
XM_017003006.1 |
XP_016858495.1
|
4, 5 |
1687 |
|
XM_017003007.1 |
XP_016858496.1
|
4, 5 |
1214 |
|
GenBank mRNA |
BC113742.1 |
- |
3, 4 |
1541 |
|
D26361.1 |
- |
4, 5 |
1902 |
|
View Full Product Details