Gene Transcripts

Gene Symbol
ZNF837
Entrez Gene ID
116412
Gene Name
zinc finger protein 837
Gene Aliases
Chromosome Location
Chr. 19: 58367623 - 58381022 on Build GRCh38
UniGene ID
Hs.222236
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138466.1 NP_612475.1 2, 3 200
GenBank mRNA BC012365.1 - 1, 2 62
BX374687.2 - 2, 3 204

View Full Product Details