Gene Transcripts
- Gene Symbol
- LOC100508689
- Entrez Gene ID
- 100508689
- Gene Name
- mucin
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L07518.1 |
- |
2 |
1044 |
|
- Gene Symbol
- MUC6
- Entrez Gene ID
- 4588
- Gene Name
- mucin 6, oligomeric mucus/gel-forming
- Gene Aliases
- MUC-6
- Chromosome Location
- Chr. 11: 1012824 - 1036706 on Build GRCh38
- UniGene ID
- Hs.528432
- Species
- Homo sapiens
View Full Product Details