Gene Transcripts
- Gene Symbol
- AAK1
- Entrez Gene ID
- 22848
- Gene Name
- AP2 associated kinase 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 69457995 - 69643845 on Build GRCh38
- UniGene ID
- Hs.468878
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014911.3 |
NP_055726.3
|
22 |
18817 |
|
GenBank mRNA |
BX640963.1 |
- |
2 |
2496 |
|
View Full Product Details