Gene Transcripts

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
Chromosome Location
Chr. 11: 94021212 - 94114208 on Build GRCh38
UniGene ID
Hs.669645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098672.1 NP_001092142.1 12 2352
GenBank mRNA BX641008.1 - 1 107

View Full Product Details