Gene Transcripts

Gene Symbol
APOL6
Entrez Gene ID
80830
Gene Name
apolipoprotein L6
Gene Aliases
APOL-VI, APOLVI
Chromosome Location
Chr. 22: 35648317 - 35668409 on Build GRCh38
UniGene ID
Hs.257352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030641.3 NP_085144.1 3 8436
XM_011530392.2 XP_011528694.1 4 8613
GenBank mRNA AK097266.1 - 1 548

View Full Product Details