Gene Transcripts

Gene Symbol
MUC5AC
Entrez Gene ID
4586
Gene Name
mucin 5AC, oligomeric mucus/gel-forming
Gene Aliases
MUC5, TBM, leB, mucin
Chromosome Location
Chr. 11: 1157953 - 1201138 on Build GRCh38
UniGene ID
Hs.534332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304359.1 NP_001291288.1 1643
GenBank mRNA AF015521.1 - 14 1643
AF043909.1 - 14 1634
AJ298317.1 - 14 1634

View Full Product Details