Gene Transcripts

Gene Symbol
STAT3
Entrez Gene ID
6774
Gene Name
signal transducer and activator of transcription 3
Gene Aliases
ADMIO, ADMIO1, APRF, HIES
Chromosome Location
Chr. 17: 42313324 - 42388505 on Build GRCh38
UniGene ID
Hs.463059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003150.3 NP_003141.2 3 471
NM_139276.2 NP_644805.1 3 493
NM_213662.1 NP_998827.1 3 385
XM_005257616.3 XP_005257673.2 3 484
XM_005257617.3 XP_005257674.2 3 479
XM_011525145.2 XP_011523447.1 3 484
XM_011525146.2 XP_011523448.1 3 484
XM_017024972.1 XP_016880461.1 3 479
XM_017024973.1 XP_016880462.1 3 484
XM_017024974.1 XP_016880463.1 3 479
XM_017024975.1 XP_016880464.1 3 472
XM_017024976.1 XP_016880465.1 3 472
GenBank mRNA AB451232.1 - 2 253
AJ012463.1 - 2 253
AK291933.1 - 3 566
AK297994.1 - 2 278
BC000627.2 - 3 390 3347434
BC014482.1 - 3 385 4909141
BI461226.1 - 3 498
L29277.1 - 3 473

View Full Product Details