Gene Transcripts

Gene Symbol
CNTN4
Entrez Gene ID
152330
Gene Name
contactin 4
Gene Aliases
AXCAM, BIG-2
Chromosome Location
Chr. 3: 2098771 - 3059080 on Build GRCh38
UniGene ID
Hs.298705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206955.1 NP_001193884.1 14 1929
NM_001206956.1 NP_001193885.1 6 651
NM_175607.2 NP_783200.1 15 1884
NM_175613.2 NP_783302.1 6 651
XM_006713004.3 XP_006713067.1 14 1897
XM_011533425.2 XP_011531727.1 14 1897
XM_011533426.2 XP_011531728.1 15 2116
XM_011533427.2 XP_011531729.1 15 2097
XM_011533428.2 XP_011531730.1 13 2086
XM_011533429.2 XP_011531731.1 13 2021
XM_011533430.2 XP_011531732.1 13 1982
XM_011533431.2 XP_011531733.1 12 4731
XM_017005782.1 XP_016861271.1 15 1839
XM_017005783.1 XP_016861272.1 16 2713
XM_017005784.1 XP_016861273.1 14 2174
XM_017005785.1 XP_016861274.1 15 1824
XM_017005786.1 XP_016861275.1 13 2086
XM_017005787.1 XP_016861276.1 13 1982
XM_017005788.1 XP_016861277.1 13 1695
GenBank mRNA AF549455.1 - 12 1927
AK057278.1 - 6 629
AK095729.1 - 13 1742
AK097633.1 - 9 1175
AK125460.1 - 14 2075
AK314396.1 - 6 629
AY090737.1 - 14 1766
BC026119.1 - 6 634 4826091

View Full Product Details