Gene Transcripts
- Gene Symbol
- CSF1
- Entrez Gene ID
- 1435
- Gene Name
- colony stimulating factor 1
- Gene Aliases
- CSF-1, MCSF
- Chromosome Location
- Chr. 1: 109910506 - 109930994 on Build GRCh38
- UniGene ID
- Hs.173894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000757.5 |
NP_000748.3
|
2, 3 |
585 |
|
NM_172210.2 |
NP_757349.1
|
2, 3 |
585 |
|
NM_172211.3 |
NP_757350.1
|
2, 3 |
585 |
|
NM_172212.2 |
NP_757351.1
|
2, 3 |
585 |
|
XM_017000369.1 |
XP_016855858.1
|
2, 3 |
260 |
|
GenBank mRNA |
AB208952.1 |
- |
2, 3 |
334 |
|
AK291660.1 |
- |
2, 3 |
347 |
|
BC021117.1 |
- |
2, 3 |
361 |
4586312 |
DB287787.1 |
- |
2, 3 |
518 |
|
M27087.1 |
- |
2, 3 |
296 |
|
M37435.1 |
- |
1, 2 |
277 |
|
M64592.1 |
- |
2, 3 |
172 |
|
M76453.1 |
- |
1, 2 |
136 |
|
U22386.1 |
- |
2, 3 |
216 |
|
X05825.1 |
- |
1, 2 |
120 |
|
View Full Product Details