Gene Transcripts

Gene Symbol
HSD17B12
Entrez Gene ID
51144
Gene Name
hydroxysteroid 17-beta dehydrogenase 12
Gene Aliases
KAR, SDR12C1
Chromosome Location
Chr. 11: 43680593 - 43856619 on Build GRCh38
UniGene ID
Hs.132513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016142.2 NP_057226.1 3, 4 510
XM_011520156.1 XP_011518458.1 3, 4 504
XM_017017881.1 XP_016873370.1 3, 4 190
GenBank mRNA AF078850.1 - 3, 4 348
AK074952.1 - 3, 4 362
AK075216.1 - 3, 4 371
AK222881.1 - 3, 4 332
AK292625.1 - 3, 4 395
AK301663.1 - 4, 5 301
BC012043.1 - 3, 4 333 3622879

View Full Product Details