Gene Transcripts

Gene Symbol
LBX2
Entrez Gene ID
85474
Gene Name
ladybird homeobox 2
Gene Aliases
LP3727
Chromosome Location
Chr. 2: 74497517 - 74503316 on Build GRCh38
UniGene ID
Hs.516124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009812.1 NP_001009812.1 2 1407
NM_001282430.1 NP_001269359.1 2 983
GenBank mRNA AY203953.1 - 2 1142
AY305861.1 - 2 1407

View Full Product Details