Gene Transcripts

Gene Symbol
PRELP
Entrez Gene ID
5549
Gene Name
proline and arginine rich end leucine rich repeat protein
Gene Aliases
MST161, MSTP161, SLRR2A
Chromosome Location
Chr. 1: 203475755 - 203491352 on Build GRCh38
UniGene ID
Hs.632481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002725.3 NP_002716.1 3 3915
NM_201348.1 NP_958505.1 3 3905
GenBank mRNA AK098245.1 - 1 212

View Full Product Details