Gene Transcripts

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
Chromosome Location
Chr. 2: 202773150 - 202871985 on Build GRCh38
UniGene ID
Hs.516629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288622.1 NP_001275551.1 2 479
NM_001288623.1 NP_001275552.1 2 424
NM_001288624.1 NP_001275553.1 1 330
NM_138468.5 NP_612477.3 3 606
GenBank mRNA AB053316.1 - 4 482
AK055978.1 - 2 221
AK092310.1 - 2 479
AL833904.1 - 3 263
BC000993.2 - 1 330
BC063876.1 - 2 137 30377894
BC093902.1 - 2 141
BC112235.1 - 2 141
DB097704.1 - 4 342

View Full Product Details