Gene Transcripts
- Gene Symbol
- IGLL5
- Entrez Gene ID
- 100423062
- Gene Name
- immunoglobulin lambda like polypeptide 5
- Gene Aliases
- IGL, IGLV, VL-MAR
- Chromosome Location
- Chr. 22: 22887780 - 22895834 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
- Gene Symbol
- CYAT1
- Entrez Gene ID
- 100290481
- Gene Name
- immunoglobulin lambda light chain-like
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB369252.1 |
- |
4 |
641 |
|
BC093098.1 |
- |
4 |
698 |
30325463 |
- Gene Symbol
- IGLC1
- Entrez Gene ID
- 3537
- Gene Name
- immunoglobulin lambda constant 1
- Gene Aliases
- IGLC
- Chromosome Location
- Chr. 22: 22895375 - 22895694 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC007782.2 |
- |
4 |
659 |
|
BC012159.1 |
- |
2 |
683 |
4575311 |
- Gene Symbol
- IGLJ3
- Entrez Gene ID
- 28831
- Gene Name
- immunoglobulin lambda joining 3
- Gene Aliases
- J3
- Chromosome Location
- Chr. 22: 22904988 - 22905025 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U07991.1 |
- |
3 |
599 |
|
X57812.1 |
- |
4 |
653 |
|
X57817.1 |
- |
4 |
644 |
|
X57821.1 |
- |
4 |
646 |
|
- Gene Symbol
- IGLV1-44
- Entrez Gene ID
- 28823
- Gene Name
- immunoglobulin lambda variable 1-44
- Gene Aliases
- IGLV144, V1-16
- Chromosome Location
- Chr. 22: 22380880 - 22381347 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC030984.1 |
- |
4 |
699 |
|
- Gene Symbol
- IGLV3-21
- Entrez Gene ID
- 28796
- Gene Name
- immunoglobulin lambda variable 3-21
- Gene Aliases
- IGLV321, V2-14
- Chromosome Location
- Chr. 22: 22712419 - 22713199 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070352.1 |
- |
4 |
684 |
|
- Gene Symbol
- IGLV3-25
- Entrez Gene ID
- 28793
- Gene Name
- immunoglobulin lambda variable 3-25
- Gene Aliases
- IGLV325, V2-17
- Chromosome Location
- Chr. 22: 22686769 - 22687281 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022823.1 |
- |
4 |
681 |
|
BC089418.1 |
- |
5 |
656 |
|
- Gene Symbol
- IGLV4-3
- Entrez Gene ID
- 28786
- Gene Name
- immunoglobulin lambda variable 4-3
- Gene Aliases
- IGLV43, V5-1
- Chromosome Location
- Chr. 22: 22871539 - 22872035 on Build GRCh38
- UniGene ID
- Hs.449585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC020236.1 |
- |
4 |
697 |
|
View Full Product Details