Gene Transcripts
- Gene Symbol
- POU5F1P4
- Entrez Gene ID
- 645682
- Gene Name
- POU class 5 homeobox 1 pseudogene 4
- Gene Aliases
- Oct4-pg4, POU5FLC1
- Chromosome Location
- Chr. 1: 155433180 - 155434262 on Build GRCh38
- UniGene ID
- Hs.632482
- Species
- Homo sapiens
- Gene Symbol
- POU5F1B
- Entrez Gene ID
- 5462
- Gene Name
- POU class 5 homeobox 1B
- Gene Aliases
- OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
- Chromosome Location
- Chr. 8: 127415612 - 127417210 on Build GRCh38
- UniGene ID
- Hs.450254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
GU480844.1 |
- |
1 |
871 |
|
HQ122875.1 |
- |
1 |
868 |
|
- Gene Symbol
- POU5F1P3
- Entrez Gene ID
- 642559
- Gene Name
- POU class 5 homeobox 1 pseudogene 3
- Gene Aliases
- OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
- Chromosome Location
- Chr. 12: 8133769 - 8134852 on Build GRCh38
- UniGene ID
- Hs.646545
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
GU480891.1 |
- |
1 |
870 |
|
View Full Product Details